Antimony(III) iodide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35676-03
35676-14
CAAA35676-03EA
83.42
CAD
CAAA35676-03
CAAA35676-14
Antimony(III) iodide, ultra dry ≥99.998% (metals basis)
Antimony (III) iodide
MDL: MFCD00016322
Decomposed by water to SbOI.
Purity: 99.998%
CAS No.: 7790-44-5
Molecular Formula: SbI3
Formula Weight: 502.46
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Moisture Sensitive
Boiling Point: 420°
Solubility: Decomposed by water to SbOI.
Merck Reference: 14,710
UN No.: 3260
MDL No.: MFCD00016322
Formula:
I₃Sb Boiling Pt: 420 °C Melting Pt: 168 °C Density: 4.676 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016322 CAS Number: 7790-44-5 EINECS: 232-205-8 UN: 3260 ADR: 8,II Merck Index: 14,710 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...