Lead titanate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead titanium oxide, Lead(II) titanium oxide, Lead(II) titanate, Lead titanate (IV), Lead titanate

35671-18 35671-30
CAAA35671-18EA 145.68 CAD
CAAA35671-18 CAAA35671-30
Lead titanate ≥99.9% (metals basis)
Lead titanate

MDL: MFCD00011159


Purity: 99.9% CAS No.: 12060-00-3 Molecular Formula: PbTiO3 Formula Weight: 303.09 Physical Form: -200 Mesh Powder Density: 7.520 Appearance: Yellow Odor: Odorless UN No.: 3288 MDL No.: MFCD00011159

Formula: O₃PbTi
MW: 303.08 g/mol
Melting Pt: 1120 °C
Density: 7,520
Storage Temperature: Ambient
MDL Number: MFCD00011159
CAS Number: 12060-00-3
EINECS: 235-038-9
UN: 3288
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR