Hafnium dioxide ≥99.95% (metals basis excluding Zr) (max. 0.5% Zr)

Supplier: Thermo Scientific Chemicals

35666-03 35666-06 35666-14
CAAA35666-03EA 70.63 CAD
CAAA35666-03 CAAA35666-06 CAAA35666-14
Hafnium dioxide ≥99.95% (metals basis excluding Zr) (max. 0.5% Zr)
Hafnium dioxide

MDL: MFCD00003565


Purity: 99.95%, CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Melting Point: 2774° Merck Reference: 14,4588 MDL No.: MFCD00003565

Formula: HfO₂
MW: 210.49
Boiling Pt: 5400
Melting Pt: 2812
Density: 9.68
Storage Temperature: Ambient
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR