Zinc phosphide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trizinc diphosphide, Zinc phosphide
35664-02
35664-04
35664-09
CAAA35664-02EA
164.32
CAD
CAAA35664-02
CAAA35664-04
CAAA35664-09
Zinc phosphide ≥99.9% (metals basis)
Zinc phosphide
MDL: MFCD00049631
Purity: 99.9%
CAS No.: 1314-84-7
Molecular Formula: P2Zn3
Formula Weight: 258.10
Physical Form: Agglomerated Powder, 3mm & Down
Density: 4.55
Appearance: Gray
Odor: Characteristic
Melting Point: 420°
Boiling Point: 1100°
Merck Reference: 14,10152
UN No.: 1714
MDL No.: MFCD00049631
Formula:
P₂Zn₃ MW: 258.12 Boiling Pt: 1100 °C Melting Pt: 420 °C Density: 4.54 Storage Temperature: Ambient |
MDL Number:
MFCD00049631 CAS Number: 1314-84-7 EINECS: 215-244-5 UN: 1714 ADR: 4.3 I Merck Index: 14,10152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...