Dipraseodymium trioxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
35663-09
35663-18
CAAA35663-09EA
199.08
CAD
CAAA35663-09
CAAA35663-18
Dipraseodymium trioxide ≥99.9% (metals basis)
Dipraseodymium trioxide
MDL: MFCD00011178
Notes: For vacuum deposition
Purity: 99.9%
CAS No.: 12036-32-7
Molecular Formula: Pr2O3
Formula Weight: 329.83
Physical Form: 3-12mm Sintered Lump
Density: 7.07
Melting Point: 2300°
Merck Reference: 14,7712
MDL No.: MFCD00011178
Formula:
O₃Pr₂ Melting Pt: 2300 °C Density: 7.07 Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS Number: 12036-32-7 EINECS: 234-857-9 Merck Index: 14,7712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...