Silicon ≥99.999% (metals basis), crystalline powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
35662-09
35662-18
CAAA35662-09EA
143.15
CAD
CAAA35662-09
CAAA35662-18
Silicon ≥99.999% (metals basis), crystalline powder -325 mesh
Silicon
MDL: MFCD00085311
Purity: 99.999%
CAS No.: 7440-21-3
Appearance: White
Odor: Odorless
UN No.: 1346
MDL No.: MFCD00085311
Formula:
Si MW: 28.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1 III Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...