Nickel sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

35661-09 35661-18
CAAAA35661-09EA 262.28 CAD
CAAAA35661-09 CAAA35661-18 CAAA35661-30
Nickel sulfide ≥99.9% (metals basis)
Nickel sulfide

MDL: MFCD00016266


Purity: 99.9% CAS No.: 12035-72-2 Molecular Formula: Ni3S2 Formula Weight: 240.19 Physical Form: -200 Mesh Powder Appearance: Black Melting Point: 797° UN No.: 3077 MDL No.: MFCD00016266

Formula: NiS
Melting Pt: 797 °C
Storage Temperature: Ambient
MDL Number: MFCD00016266
CAS Number: 16812-54-7
EINECS: 234-829-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Form -200 Mesh Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR