Cerium(IV) sulfate standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
35650-K2
35650-K7
CAAA35650-K2EA
178.79
CAD
CAAA35650-K2
CAAA35650-K7
Cerium(IV) sulfate standardised solution 0.1 N in aqueous solution
Cerium (IV) sulphate
MDL: MFCD00148852
CAS No.: 13590-82-4
Molecular Formula: CeO8S2
Formula Weight: 332.24
Physical Form: Liquid
Density: 1.05
Appearance: Orange
Merck Reference: 14,1990
UN No.: 3264
MDL No.: MFCD00148852
Formula:
Ce(SO₄)₂ Density: 1.05 Storage Temperature: Ambient |
MDL Number:
MFCD00148852 CAS Number: 13590-82-4 EINECS: 237-029-5 UN: 3264 ADR: 8,III Merck Index: 14,1990 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...