Hydrochloric acid standardised solution 0.01 N

Supplier: Thermo Scientific Chemicals

35648-K2 35648-K7 35648-M4
CAAA35648-K2EA 71.97 CAD
CAAA35648-K2 CAAA35648-K7 CAAA35648-M4
Hydrochloric acid standardised solution 0.01 N
Hydrochloric acid

MDL: MFCD00011324


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless

Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR