Hydrochloric acid standardised solution 0.05 N

Supplier: Thermo Scientific Chemicals

Synonyms: Hydrochloric acid solution, Muriatic acid

35646-K2 35646-K7 35646-M4
CAAA35646-K2EA 70.82 CAD
CAAA35646-K2 CAAA35646-K7 CAAA35646-M4
Hydrochloric acid standardised solution 0.05 N
Hydrochloric acid

MDL: MFCD00011324


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless

Formula: HCl
MW: 36.46
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR