Sodium thiosulfate standardised solution 0.01 N in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

35643-K2 35643-K7 35643-M4
CAAA35643-K2EA 75.18 CAD
CAAA35643-K2 CAAA35643-K7 CAAA35643-M4
Sodium thiosulfate standardised solution 0.01 N in aqueous solution
Sodium thiosulphate

MDL: MFCD00003499


CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid MDL No.: MFCD00003499

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR