Hydrochloric acid standardised solution 1.0 N

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

35640-K2 35640-K7 35640-M4
CAAA35640-K2EA 83.9 CAD
CAAA35640-K2 CAAA35640-K7 CAAA35640-M4
Hydrochloric acid standardised solution 1.0 N
Hydrochloric acid

MDL: MFCD00011324


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless to pale yellow

Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR