Sodium hydroxide 40% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
35639-K7
35639-M4
CAAA35639-K7EA
173.8
CAD
CAAA35639-K7
CAAA35639-M4
Sodium hydroxide 40% (w/v) in aqueous solution
Sodium hydroxide
MDL: MFCD00003548
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
UN No.: 1824
MDL No.: MFCD00003548
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...