Hydrochloric acid standardised solution 5.0 N

Supplier: Thermo Scientific Chemicals
Danger

35638-K2 35638-K7 35638-M4
CAAA35638-K2EA 82.79 CAD
CAAA35638-K2 CAAA35638-K7 CAAA35638-M4
Hydrochloric acid standardised solution 5.0 N
Hydrochloric acid

MDL: MFCD00011324


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless to pale yellow UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR