Sodium hydroxide standardised solution 5.0 N in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
35633-K2
35633-K7
CAAA35633-K2EA
90.53
CAD
CAAA35633-K2
CAAA35633-K7
Sodium hydroxide standardised solution 5.0 N in aqueous solution
Sodium hydroxide
MDL: MFCD00003548
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
UN No.: 1824
MDL No.: MFCD00003548
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...