Sodium hydroxide standardised solution 0.5 N in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
35627-K2
35627-K7
35627-M4
CAAA35627-K2EA
58.93
CAD
CAAA35627-K2
CAAA35627-K7
CAAA35627-M4
Sodium hydroxide standardised solution 0.5 N in aqueous solution
Sodium hydroxide
MDL: MFCD00003548
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
Odor: Odorless
UN No.: 3266
MDL No.: MFCD00003548
Formula:
NaOH Boiling Pt: ∼105 °C (1013 hPa) Density: 1.02 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...