Nitric acid standardised solution 1.0 N

Supplier: Thermo Scientific Chemicals
Danger

35624-AP 35624-K2
CAAA35624-APEA 76.47 CAD
CAAA35624-AP CAAA35624-K2
Nitric acid standardised solution 1.0 N
Nitric acid

MDL: MFCD00011349


CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colorless UN No.: 3264 MDL No.: MFCD00011349

Formula: HNO₃
Density: 1.035 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR