Potassium hydroxide 50% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Caustic potash

35621-AP 35621-K2 35621-K7
CAAA35621-APEA 85.32 CAD
CAAA35621-AP CAAA35621-K2 CAAA35621-K7
Potassium hydroxide 50% (w/v) in aqueous solution
Potassium hydroxide

MDL: MFCD00003553


CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid Density: 1.50 Appearance: Colorless UN No.: 1814 MDL No.: MFCD00003553

Formula: KOH
Density: 1.50
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR