Sodium hydroxide standardised solution 0.01 N in aqueous solution

Supplier: Thermo Scientific Chemicals

35620-K2 35620-K7 35620-M4
CAAA35620-K2EA 63.25 CAD
CAAA35620-K2 CAAA35620-K7 CAAA35620-M4
Sodium hydroxide standardised solution 0.01 N in aqueous solution
Sodium hydroxide

MDL: MFCD00003548


CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00003548

Formula: NaOH
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR