Oxalic acid 5% w/v aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanedioic acid
35619-K2
35619-K7
CAAA35619-K2EA
81.37
CAD
CAAA35619-K2
CAAA35619-K7
Oxalic acid 5% w/v aqueous solution
Oxalic acid
MDL: MFCD00002573
Molecular Formula: HO2CCO2H
Formula Weight: 90.04
Physical Form: Liquid
Odor: Odorless
UN No.: 3265
MDL No.: MFCD00002573
Formula:
C₂H₂O₄ MW: 90.04 g/mol Boiling Pt: 365 Storage Temperature: Ambient |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 UN: 3265 ADR: 8 III Merck Index: 13,06980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...