Potassium dichromate standardised solution 0.1 N

Supplier: Thermo Scientific Chemicals
Danger

35618-AP 35618-K2
CAAA35618-APEA 53.72 CAD
CAAA35618-AP CAAA35618-K2
Potassium dichromate standardised solution 0.1 N
Potassium dichromate

MDL: MFCD00011367


CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid Appearance: Orange Odor: Odorless MDL No.: MFCD00011367

Formula: K₂Cr₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Concentration 0.1 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR