Oxalic acid 10% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

35617-K2 35617-K7
CAAA35617-K2EA 97.8 CAD
CAAA35617-K2 CAAA35617-K7
Oxalic acid 10% (w/v) in aqueous solution
Oxalic acid

MDL: MFCD00002573


Molecular Formula: HO2CCO2H Formula Weight: 90.04 Physical Form: Liquid Odor: Odorless UN No.: 3265 MDL No.: MFCD00002573

Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR