Sodium chloride standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals

35616-AP 35616-K2
CAAA35616-APEA 60.83 CAD
CAAA35616-AP CAAA35616-K2
Sodium chloride standardised solution 0.1 N in aqueous solution
Sodium chloride

MDL: MFCD00003477


CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: Liquid MDL No.: MFCD00003477

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR