Hydrochloric acid 10% (v/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

35611-AP 35611-K2 35611-K7
CAAA35611-APEA 46.93 CAD
CAAA35611-AP CAAA35611-K2 CAAA35611-K7
Hydrochloric acid 10% (v/v) in aqueous solution
Hydrochloric acid

MDL: MFCD00011324


CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless to pale yellow UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.02…1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR