Perchloric acid standardised solution 0.1 N in acetic acid
Supplier: Thermo Scientific Chemicals
|
Danger
|
35608-K2
35608-K7
CAAA35608-K2EA
167.48
CAD
CAAA35608-K2
CAAA35608-K7
Perchloric acid standardised solution 0.1 N in acetic acid
Perchloric acid
MDL: MFCD00011325
CAS No.: 7601-90-3
Molecular Formula: HClO4
Formula Weight: 100.46
Physical Form: Liquid
Odor: Pungent
pH: 2.5
UN No.: 2920
MDL No.: MFCD00011325
Formula:
HClO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 2920 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...