Potassium thiocyanate standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals

35602-K2 35602-K7
CAAA35602-K2EA 106.81 CAD
CAAA35602-K2 CAAA35602-K7
Potassium thiocyanate standardised solution 0.1 N in aqueous solution
Potassium thiocyanate

MDL: MFCD00011413


CAS No.: 333-20-0 Molecular Formula: KSCN Formula Weight: 97.18 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00011413

Formula: KSCN
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR