Potassium hydroxide standardised solution 0.1 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

35599-K2 35599-K7 35599-M4
CAAA35599-K2EA 65.25 CAD
CAAA35599-K2 CAAA35599-K7 CAAA35599-M4
Potassium hydroxide standardised solution 0.1 N in aqueous solution
Potassium hydroxide

MDL: MFCD00003553


CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid Appearance: Colorless

Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR