Silver nitrate standardised solution 1.0 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

35597-AP 35597-K2
CAAA35597-APEA 1426.74 CAD
CAAA35597-AP CAAA35597-K2
Silver nitrate standardised solution 1.0 N in aqueous solution
Silver nitrate

MDL: MFCD00003414


CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colorless UN No.: 3264 MDL No.: MFCD00003414

Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR