Barium chloride standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Warning
|
35582-K2
35582-K7
CAAA35582-K2EA
124.19
CAD
CAAA35582-K2
CAAA35582-K7
Barium chloride standardised solution 0.1 N in aqueous solution
Barium chloride
MDL: MFCD00003445
CAS No.: 10361-37-2
Molecular Formula: BaCl2
Formula Weight: 208.25
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00003445
Formula:
BaCl₂ Density: 1.034 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003445 CAS Number: 10361-37-2 EINECS: 233-788-1 UN: 3287 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...