Ammonia solution 10% (v/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

35575-AP 35575-K2 35575-K7
CAAA35575-APEA 73 CAD
CAAA35575-AP CAAA35575-K2 CAAA35575-K7
Ammonia solution 10% (v/v) in aqueous solution
Ammonia solution

MDL: MFCD00066650


CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.98 Appearance: Colorless Odor: Ammonia-like UN No.: 2672 MDL No.: MFCD00066650

Formula: NH₃
Density: 0.98
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR