Acetic acid 4% v/v aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

35572-AP 35572-K2
CAAA35572-APEA 52.93 CAD
CAAA35572-AP CAAA35572-K2
Acetic acid 4% v/v aqueous solution
Acetic acid

MDL: MFCD00036152


Molecular Formula: CH3CO2H Formula Weight: 60.05 Physical Form: Liquid Odor: Pungent MDL No.: MFCD00036152

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR