Acetic acid 1% (v/v) in aqueous solution

Supplier: Thermo Scientific Chemicals

35569-AP 35569-K2
CAAA35569-APEA 46.77 CAD
CAAA35569-AP CAAA35569-K2
Acetic acid 1% (v/v) in aqueous solution
Acetic acid

MDL: MFCD00036152


Molecular Formula: CH3CO2H Formula Weight: 60.05 Physical Form: Liquid MDL No.: MFCD00036152

Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR