Cadmium ≥99.999% (metals basis), tear drop shot 3 mm (0.1 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35483-18
35483-30
CAAA35483-18EA
254.38
CAD
CAAA35483-18
CAAA35483-30
Cadmium ≥99.999% (metals basis), tear drop shot 3 mm (0.1 in)
Cadmium
MDL: MFCD00010914
Purity: 99.999%
CAS No.: 7440-43-9
MDL No.: MFCD00010914
Formula:
Cd Boiling Pt: 765 °C (1013 hPa) Melting Pt: 321 °C Density: 8.642 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010914 CAS Number: 7440-43-9 EINECS: 231-152-8 UN: 2570 ADR: 6.1,II Merck Index: 12,01649 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...