Trilithium nitride ≥99.4% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

35479-09 35479-18
CAAA35479-09EA 126.08 CAD
CAAA35479-09 CAAA35479-18
Trilithium nitride ≥99.4% (metals basis)
tri-Lithium nitride

MDL: MFCD00016186


Purity: 99.4% CAS No.: 26134-62-3 Molecular Formula: Li3N Formula Weight: 34.82 Physical Form: -60 Mesh Powder Density: 1.30 Appearance: Red to brown Odor: Ammonia-like Melting Point: 845° UN No.: 2806 MDL No.: MFCD00016186

Formula: Li₃N
Melting Pt: 845 °C
Density: 1.30
Storage Temperature: Ambient
MDL Number: MFCD00016186
CAS Number: 26134-62-3
EINECS: 247-475-2
UN: 2806
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR