Sodium aluminate, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium Aluminium oxide, Aluminum sodium oxide, Aluminium sodium dioxide, Sodium aluminum oxide
35453-22
35453-36
CAAA35453-22EA
108.7
CAD
CAAA35453-22
CAAA35453-36
Sodium aluminate, tech.
Sodium aluminate
MDL: MFCD00801010
CAS No.: 1302-42-7
Molecular Formula: Al2O3.Na2O
Formula Weight: 163.94
Physical Form: Crystalline
Density: 1.52
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Solubility: Insoluble in alcohol, soluble in water
UN No.: 2812
MDL No.: MFCD00801010
Formula:
NaAlO₂ Melting Pt: 1800 °C Density: 1.52 Storage Temperature: Ambient |
MDL Number:
MFCD00801010 CAS Number: 1302-42-7 EINECS: 215-100-1 UN: 2812 ADR: 8,III Merck Index: 12,08715 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...