Magnesium oxide ≥95.0% ACS
Supplier: Thermo Scientific Chemicals
35442-22
35442-36
35442-A3
CAAA35442-22EA
148.2
CAD
CAAA35442-22
CAAA35442-36
CAAA35442-A3
Magnesium oxide ≥95.0% ACS
Magnesium oxide
MDL: MFCD00011109
Purity: 95.0% min
CAS No.: 1309-48-4
Molecular Formula: MgO
Formula Weight: 40.30
Physical Form: Granular
Density: 3.58
Refractive Index: 1.736
Odor: Odorless
Melting Point: 2852°
Boiling Point: 3600°
Merck Reference: 14,5677
MDL No.: MFCD00011109
Formula:
MgO Boiling Pt: 3600 °C Melting Pt: 2852 °C Density: 3.58 Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 14,5677 |
Specification Test Results
Insoluble in dilute hydrochloric acid | 0.02% |
Soluble in water | 0.4% |
Loss on ignition | 2% |
Cl | 0.01% |
NO3 | 0.005% |
Sulfate and sulfite (as SO3) | 0.02% |
Ba | 0.005% |
Ca | 0.05% |
Heavy Metals (as Pb) | 0.003%% |
Fe | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...