Dichlorodiphenylgermane 98+%

Supplier: Thermo Scientific Chemicals
Danger

34113-03 34113-06 34113-14
CAAA34113-03EA 108.39 CAD
CAAA34113-03 CAAA34113-06 CAAA34113-14
Dichlorodiphenylgermane 98+%
Dichlorodiphenylgermane

MDL: MFCD00013587


Purity: 98+% CAS No.: 1613-66-7 Molecular Formula: (C6H5)2GeCl2 Formula Weight: 297.73 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.415 Refractive Index: 1.5970 Sensitivity: Moisture Sensitive Melting Point: 9° Boiling Point: 107°/16mm UN No.: 3265 MDL No.: MFCD00013587

Formula: C₁₂H₁₀Cl₂Ge
Boiling Pt: 107 °C (16 mmHg)
Melting Pt: 9 °C
Density: 1,415
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013587
CAS Number: 1613-66-7
EINECS: 216-562-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR