Sodium bis(trimethylsilyl)amide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
HMDS sodium salt, N-Sodiohexamethyldisilazane, Sodium bis(trimethylsilyl)amide, HMDS-Na
33505-04
33505-09
33505-18
CAAA33505-04EA
46.14
CAD
CAAA33505-04
CAAA33505-09
CAAA33505-18
Sodium bis(trimethylsilyl)amide 98%
Sodium bis(trimethylsilyl)amide
MDL: MFCD00009835
Soluble in hexane, toluene, ether
Purity: 98%
CAS No.: 1070-89-9
Molecular Formula: C6H18NNaSi2
Formula Weight: 183.38
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Odorless
Melting Point: 171-175°
Solubility: Soluble in hexane, toluene, ether
UN No.: 3096
MDL No.: MFCD00009835
Formula:
C₆H₁₈NNaSi₂ MW: 183.38 g/mol Melting Pt: 171…175 °C Density: 1.065 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009835 CAS Number: 1070-89-9 EINECS: 213-983-8 UN: 3096 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...