Osmium(III) chloride trihydrate ≥99.99% (metals basis) Os 52-56%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
33480-01
33480-03
33480-06
CAAA33480-01EA
150.57
CAD
CAAA33480-01
CAAA33480-03
CAAA33480-06
Osmium(III) chloride trihydrate ≥99.99% (metals basis) Os 52-56%, Premion®
Osmium (III) chloride trihydrate
Purity: 99.99%
CAS No.: 135296-80-9
Molecular Formula: Cl3Os.3H2O
Formula Weight: 350.60 (296.56anhy)
Physical Form: Crystalline
Appearance: Red-black
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011148
Formula:
Cl₃Os·H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011148 CAS Number: 135296-80-9 EINECS: 236-587-7 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...