Strontium chloride hexahydrate 99.0-103.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
33393-22
33393-36
CAAA33393-22EA
77.1
CAD
CAAA33393-22
CAAA33393-36
Strontium chloride hexahydrate 99.0-103.0% ACS
Strontium chloride hexahydrate
MDL: MFCD00149865
Notes: m.p. 61°C (rapidly heated) , 150°C -6H2O
Purity: 99.0-103.0%
CAS No.: 10025-70-4
Molecular Formula: SrCl2.6H2O
Formula Weight: 266.62 (158.53anhy)
Physical Form: Granular
Density: 1.93
Refractive Index: 1.650
Sensitivity: Hygroscopic
Merck Reference: 14,8840
MDL No.: MFCD00149865
Formula:
SrCl₂·6H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 61 °C Density: 1.93 Storage Temperature: Ambient |
MDL Number:
MFCD00149865 CAS Number: 10025-70-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5% solution | 5.0-7.0 at 25° |
SO3 | 0.001% |
Ba | 0.05% |
Ca | 0.05% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm |
MG | 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...