Strontium carbonate ≥97.5%

Supplier: Thermo Scientific Chemicals

33392-30 33392-A1 33392-A7
CAAA33392-30EA 63.52 CAD
CAAA33392-30 CAAA33392-A1 CAAA33392-A7
Strontium carbonate ≥97.5%
Strontium carbonate

As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnics


Purity: 97.5% CAS No.: 1633-05-2 Molecular Formula: SrCO3 Formula Weight: 147.63 Physical Form: Powder Density: 3.70 Melting Point: 1100° dec. Solubility: Practically insoluble in water. Soluble in acids and carbonated water Merck Reference: 14,8838 MDL No.: MFCD00011250


As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnics

Formula: CO₃Sr
MW: 147.63 g/mol
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1100 °C (dec.)
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,8838

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR