Sodium thiocyanate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
33388-22
33388-36
33388-A3
CAAA33388-22EA
77.89
CAD
CAAA33388-22
CAAA33388-36
CAAA33388-A3
Sodium thiocyanate ≥98.0% ACS
Sodium thiocyanate
MDL: MFCD00011123
Beilstein Registry No.: 3594965
Soluble in water, acetone, alcohol
Fieser: 1,1105
Purity: 98.0% min
CAS No.: 540-72-7
Molecular Formula: NaSCN
Formula Weight: 81.07
Physical Form: Crystalline/Chunks
Density: 1.74
Appearance: White
Odor: Odorless
pH: 6-8
Melting Point: 287°
Solubility: Soluble in water, acetone, alcohol
Beilstein Reference: 3594965
Merck Reference: 14,9327
MDL No.: MFCD00011123
Formula:
NaSCN Melting Pt: 287 °C Density: 1.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011123 CAS Number: 540-72-7 EINECS: 208-754-4 Merck Index: 14,9327 |
Specification Test Results
Insoluble matter | 0.005% |
Na2CO3 | 0.2% |
Cl | 0.01% |
SO3 | 0.01% |
S | 0.001% |
NH3 | 0.002% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...