di-Sodium succinate hexahydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate
MDL: MFCD00149117
Beilstein Registry No.: 3921044
CAS No.: 6106-21-4
Molecular Formula: C4H4Na2O4.6H2O
Formula Weight: 270.14 (162.05anhy)
Physical Form: Granular/crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 120° -6H2O
Beilstein Reference: 3921044
Merck Reference: 14,8679
MDL No.: MFCD00149117
Formula:
NaOOCCH₂CH₂COONa·6H₂O Melting Pt: 120 °C (-6H₂O) Density: 1.529 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149117 CAS Number: 6106-21-4 EINECS: 205-778-7 Merck Index: 14,8679 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...