Tetrasodium diphosphate decahydrate 99.0-103.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
33385-22
33385-36
33385-A3
CAAA33385-22EA
54.35
CAD
CAAA33385-22
CAAA33385-36
CAAA33385-A3
Tetrasodium diphosphate decahydrate 99.0-103.0% ACS
tetra-Sodium diphosphate decahydrate
MDL: MFCD00149200
Purity: 99.0-103.0%
CAS No.: 13472-36-1
Molecular Formula: Na4O7P2.10H2O
Formula Weight: 446.06 (265.90anhy)
Physical Form: Crystalline
Density: 1.82
pH: 9.5-10.5
Melting Point: 79.5°
Merck Reference: 14,9240
MDL No.: MFCD00149200
Formula:
Na₄P₂O₇·10H₂O MW: 446.06 g/mol Melting Pt: 79.5 °C Density: 1.82 Storage Temperature: Ambient |
MDL Number:
MFCD00149200 CAS Number: 13472-36-1 EINECS: 231-767-1 Merck Index: 14,9240 |
Specification Test Results
Insoluble matter | 0.01% |
pH of a 5% solution | 9.5-10.5 at 25° |
Cl | 0.002% |
SO3 | 0.005% |
N | 0.001% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...