Sodium peroxide ≥93% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Sodium peroxide
As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent
Purity: 93% min
CAS No.: 1313-60-6
Molecular Formula: Na2O2
Formula Weight: 77.98
Physical Form: Granular, Packaged in screwtop can
Density: 2.805
Appearance: White to pale yellow
Melting Point: 460° dec.
Solubility: Soluble in water, forming NaOH and H2O2
Merck Reference: 14,8655
UN No.: 1504
MDL No.: MFCD00003497
As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent
Formula:
Na₂O₂ MW: 77.98 g/mol Melting Pt: 460 °C (dec.) Density: 2.805 Storage Temperature: Ambient |
MDL Number:
MFCD00003497 CAS Number: 1313-60-6 EINECS: 215-209-4 UN: 1504 ADR: 5.1,I Merck Index: 14,8655 |
Specification Test Results
Cl | 0.002% |
PO3 | 5 ppm |
SO3 | 0.001% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.005% |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...