Sodium perborate tetrahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
33383-14
33383-22
33383-36
CAAA33383-14EA
58.62
CAD
CAAA33383-14
CAAA33383-22
CAAA33383-36
Sodium perborate tetrahydrate, REacton®, Reagent Grade
Sodium perborate tetrahydrate
Bleaching agent for domestic detergents and industrial laundries
CAS No.: 10486-00-7
Molecular Formula: NaBO3.4H2O
Formula Weight: 153.86 (81.84anhy)
Physical Form: Granular
Density: 1.73
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: ca 10
Melting Point: 60° dec.
Merck Reference: 14,8652
UN No.: 1479
MDL No.: MFCD00149231
Bleaching agent for domestic detergents and industrial laundries
Formula:
NaH₂BO₄·4H₂O Melting Pt: 60 °C (dec.) Density: 1.73 Storage Temperature: Ambient |
MDL Number:
MFCD00149231 CAS Number: 13517-20-9 EINECS: 239-172-9 UN: 1479 ADR: 5.1,III Merck Index: 14,8652 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...