Sodium carbonate 99.95-100.05% (dried basis) ACS primary standard

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

33377-22 33377-36
CAAA33377-22EA 61.46 CAD
CAAA33377-22 CAAA33377-36
Sodium carbonate 99.95-100.05% (dried basis) ACS primary standard
Sodium carbonate

MDL: MFCD00003494 Beilstein Registry No.: 4154566 Notes: m.p. 851°C (begins to lose CO2 at 400°C). For use as an alkalimetric standard, material should be heated at 285°C for 2 hrs. Fieser: 7,332


Physical Form: Granular Density: 2.53 Refractive Index: 1.535 Sensitivity: Hygroscopic Odor: Odorless pH: 11,5 Merck Reference: 14,8596

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
Loss on drying at 285°C 1%
Cl 0.001%
Nitrogen compounds (as N) 0.00001
PO3 0.005%
Sulfur compounds (as SO3) 0.003%
Ammonium hyrdoxide precipitate 0.01%
Ca 0.02%%
MG 0.004%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR