Sodium metabisulfite SO₂ ≥ 58,5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite
MDL: MFCD00167602
Freely soluble in water, glycerol. Slightly soluble in alcohol
CAS No.: 7681-57-4
Molecular Formula: Na2S2O5
Formula Weight: 190.10
Physical Form: Granular
Density: 1.480
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale yellow
Odor: Like rotten eggs
pH: 3.5-5.0
Melting Point:
Formula:
Na₂S₂O₅ MW: 190.11 g/mol Melting Pt: 150 Density: 1.480 Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 EINECS: 231-673-0 REACH: 01-2119531326-45 Merck Index: 14,8638 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...