Disodium hydrogenarsenate heptahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
33373-14
33373-22
CAAA33373-14EA
249.64
CAD
CAAA33373-14
CAAA33373-22
Disodium hydrogenarsenate heptahydrate 98.0-102.0% ACS
Disodium hydrogenarsenate heptahydrate
MDL: MFCD00149179
Purity: 98.0-102.0%
CAS No.: 10048-95-0
Molecular Formula: HAsNa2O4.7H2O
Formula Weight: 312.01 (185.94anhy)
Physical Form: Crystalline powder
Density: 1.88
Appearance: White
Melting Point: 130° dec.
UN No.: 1685
MDL No.: MFCD00149179
Formula:
HAsNa₂O₄·7H₂O Melting Pt: 130 °C (dec.) Density: 1.88 Storage Temperature: Ambient |
MDL Number:
MFCD00149179 CAS Number: 10048-95-0 EINECS: 231-902-4 UN: 1685 ADR: 6.1,II |
Specification Test Results
Insoluble matter | 0.005% |
As2O3 | 0.01% |
Cl | 0.001% |
NO3 | 0.005% |
SO3 | 0.01% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...