Dipotassium hydrogen orthophosphate trihydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

33365-30 33365-A1
CAAA33365-30EA 105.07 CAD
CAAA33365-30 CAAA33365-A1
Dipotassium hydrogen orthophosphate trihydrate, REacton®, Reagent Grade
di-Potassium hydrogen orthophosphate trihydrate

MDL: MFCD00149926


CAS No.: 16788-57-1 Molecular Formula: K2HPO4.3H2O Formula Weight: 228.23 (174.18anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,7658 MDL No.: MFCD00149926

Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5
Merck Index: 14,7658

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR